missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, puriss. p.a., Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 6 x 1 kg |
Linear Formula | KCl |
UN Number | NONH for all modes of transport |
IUPAC Name | potassium;chloride |
Formula Weight | 74.55 |
Percent Purity | 99.5 to 100.5% |
Grade | puriss. p.a. |
Density | 1.98g/mL at 25°C |
Chemical Name or Material | Potassium Chloride, Reag. Ph. Eur. reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'