missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, MP Biomedicals™

Product Code. 11430552 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430552 500 g 500g
11420552 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430552 Supplier MP Biomedicals Supplier No. 02191427.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the preparation of phosphate buffered saline (PBS) and for the extraction and solubilization of proteins. Potassium Chloride, MP Biomedicals is a good potassium source widely used as a reagent in biochemistry and molecular biology.

Widely used reagent in biochemistry and molecular biology:

  • In the preparation of phosphate buffered saline (PBS)
  • For the extraction and solubilization of proteins
  • In the hypotonic KCl-propidium iodide protocol for univariate analysis of metaphase chromosomes
  • For dissociation of eukaryotic ribosomes into subunits by high concentrations of potassium (0.3-1.0 M)
  • In the preparation of mitochondria, provides an ionic osmotic support for those tissues that assume a gelatinous consistency upon homogenization
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water (1g/3mL, clear, colorless solution), boiling water (1g/1.8mL), glycerol (1g/14mL), or ethanol (1g/250mL); insoluble in ether or acetone; hydrochloric acid, sodium or magnesium chlorides diminish its solubility in water.
Formula Weight 74.55
Percent Purity 99.0 to 100.5%
Grade ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.