missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407726
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
4873
CHEBI:32588
[Cl-].[K+]
This item is not returnable. View return policy
7447-40-7
ClK
74.55
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
4873
CHEBI:32588
[Cl-].[K+]

The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia with its pharmaceutical preparation, a condition in which the level of potassium will be low in blood. Further it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1500°C (sublimation)
Melting Point 773°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.995% (metals basis)
Solubility Information Freely soluble in water. Slightly soluble in alcohol. Insoluble in ether.
IUPAC Name potassium;chloride
Formula Weight 74.55
Odor Odorless
Refractive Index 1.49
Sensitivity Hygroscopic
Density 1.984 g/cm3 at 20°C
Chemical Name or Material Potassium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'