missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 0.005M, Conductivity Standard, Thermo Scientific Chemicals

Product Code. 11316139
Click to view available options
Quantity:
500 mL
1 L
4 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium chloride 0.0005M solution is used as conductive standard and as electrolyte in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Concentration 0.005M
Quantity 500 mL
Formula Weight 74.55
Odor Odorless
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Potassium chloride Conductivity Standard, 718μmho at 25°C

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'