missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chlorate, puriss., 99 to 101%, Honeywell Fluka™

Product Code. 15681160 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15681160 5 kg Poly Bottle 5kg
15671160 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15681160 Supplier Honeywell-Fluka Supplier No. 126345KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Puriss., 99-101%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
MDL Number MFCD00011361
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
PubChem CID 6426889
IUPAC Name potassium;chlorate
SMILES [O-]Cl(=O)=O.[K+]

Specifications

Melting Point 356°C
Color White
pH 5 to 6.5
Physical Form Solid
Quantity 5 kg
Linear Formula KClO3
UN Number UN1485
Formula Weight 122.55g/mol
Percent Purity 99 to 101%
Grade puriss.
Packaging Poly Bottle
Appearance Crystals
Density 2.34g/cu.cm
Chemical Name or Material Potassium Chlorate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.