missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chlorate, ≥99%, ACS Reagent, Honeywell Fluka™

Product Code. 15672750 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15672750 100 g Poly Bottle 100g
15682750 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15672750 Supplier Honeywell-Fluka Supplier No. 255572100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
MDL Number MFCD00011361
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
ChEBI CHEBI:81951

Specifications

Melting Point 356°C
Color White
Quantity 100 g
Linear Formula KClO3
UN Number UN1485
Synonym benazolin, metizolin, eunasin, cornox cwk, benazalox, benopan, bensecal, cresopur, galipan, herbazolin
SMILES [K+].[O-][Cl](=O)=O
IUPAC Name potassium chlorate
PubChem CID 19662
Formula Weight 122.55g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystalline
Density 2.34g/cm3
Chemical Name or Material Potassium Chlorate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.