missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium channel subfamily K member 6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-34063
This item is not returnable.
View return policy
Description
Potassium channel subfamily K member 6 Polyclonal specifically detects Potassium channel subfamily K member 6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Potassium channel subfamily K member 6 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q9Y257 | |
| KCNK6 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LQTFRHVSDLHGLTELILLPPPCPASFNADEDDRVDILGPQPESHQQLSASSHTDY | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Inward rectifying potassium channel protein TWIK-2, K2p6.1, K2P6.1 potassium channel, potassium channel subfamily K member 6, potassium channel, subfamily K, member 6, TOSSFLJ12282, TWIK-2, TWIK2KCNK8, TWIK-originated similarity sequence, TWIK-originated sodium similarity sequence | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9424 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction