missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium channel subfamily K member 6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93795-0.1ml
This item is not returnable.
View return policy
Description
Potassium channel subfamily K member 6 Polyclonal antibody specifically detects Potassium channel subfamily K member 6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Potassium channel subfamily K member 6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Inward rectifying potassium channel protein TWIK-2, K2p6.1, K2P6.1 potassium channel, potassium channel subfamily K member 6, potassium channel, subfamily K, member 6, TOSSFLJ12282, TWIK-2, TWIK2KCNK8, TWIK-originated similarity sequence, TWIK-originated sodium similarity sequence | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 254-313 of human KCNK6 (NP_004814.1). QTFRHVSDLHGLTELILLPPPCPASFNADEDDRVDILGPQPESHQQLSASSHTDYASIPR | |
| 0.1 mL | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 9424 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction