missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Channel Kv3.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-30395
This item is not returnable.
View return policy
Description
Potassium Channel Kv3.1 Polyclonal specifically detects Potassium Channel Kv3.1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Potassium Channel Kv3.1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| P48547 | |
| KCNC1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EEALDSFGGAPLDNSADDADADGPGDSGDGEDELEMTKRLALSDS | |
| 0.1 mL | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission, Potassium Channels | |
| 3746 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ41162, FLJ42249, FLJ43491, KV3.1, KV4, MGC129855, NGK2, potassium voltage-gated channel subfamily C member 1, potassium voltage-gated channel, Shaw-related subfamily, member 1, voltage-gated potassium channel protein KV3.1, Voltage-gated potassium channel subunit Kv3.1, Voltage-gated potassium channel subunit Kv4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction