missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate Sesquihydrate, ACS Reagent, 99%, Honeywell™ Fluka™

Product Code. 15692620 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15692620 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15692620 Supplier Honeywell-Fluka Supplier No. 2435581KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6381-79-9
Color Colorless
Physical Form Crystalline
Quantity 1 kg
Molecular Formula C2H6K4O9
Linear Formula K2CO3 · 1.5H2O
MDL Number MFCD00149925
UN Number NONH for all modes of transport
InChI Key WWVGGBSGYNQKCY-UHFFFAOYSA-J
SMILES O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O
IUPAC Name tetrapotassium trihydrate dicarbonate
Molecular Weight (g/mol) 330.45
Formula Weight 165.23g/mol
Percent Purity 99%
Grade ACS Reagent
Packaging Poly bottle
Chemical Name or Material Potassium Carbonate Sesquihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.