missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate sesquihydrate, ACS, 98.5-101.0%

Product Code. 11391568
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391568 100 g 100g
11301578 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391568 Supplier Thermo Scientific Alfa Aesar Supplier No. 010981.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Manufacturing of Soaps, Pigments, Printing Ink | CAS: 6381-79-9 | C2H6K4O9 | 330.45 g/mol

Potassium carbonate sesquihydrate is used for manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts and as an analytical lab reagent. It is also useful as a reagent for removing carbon dioxide from ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-79-9
Molecular Formula C2H6K4O9
Molecular Weight (g/mol) 330.45
MDL Number MFCD00149925
InChI Key WWVGGBSGYNQKCY-UHFFFAOYSA-J
IUPAC Name tetrapotassium trihydrate dicarbonate
SMILES O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98.5-101.0%
Merck Index 14,7670
Solubility Information Soluble in water; Practically insoluble in alcohol
Formula Weight 165.23 (138.21 Anhydrous)
Percent Purity 98.5 to 101.0%
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Potassium carbonate sesquihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.