missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium carbonate BioXtra, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
Description
- Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
- Includes application-specific formulations, such as tablets and prepared solutions.
Specifications
Quantity | 1 kg |
Linear Formula | K2CO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4267587 |
IUPAC Name | dipotassium;carbonate |
Formula Weight | 138.21g/mol |
Percent Purity | ≥99% |
Chemical Name or Material | Potassium carbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'