missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate Anhydrous (White Granular Powder), Fisher BioReagents

Product Code. 10687623
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10687623 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10687623 Supplier Fisher BioReagents Supplier No. BP365500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 584-08-7 | CK2O3 | 138.21 g/mol

Glass Bottle; 500g

This potassium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Carbonate Anhydrous
Melting Point 891°C
CAS 584-08-7
Color White
pH 11.5 to 12
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥99 %
Molecular Formula CK2O3
MDL Number MFCD00011382
Merck Index 15, 7740
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES [K+].[K+].[O-]C([O-])=O
IUPAC Name dipotassium;carbonate
Molecular Weight (g/mol) 138.21
PubChem CID 11430
Formula Weight 138.21
Percent Purity ≥99%
Identification Pass Test
Packaging Glass Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.