missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, ACS, 99.0% min

Product Code. 11347457
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
25g
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347457 5 kg 5kg
11367457 25 g 25g
11327457 100 g 100g
11357457 500 g 500g
11337457 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11347457 Supplier Thermo Scientific Alfa Aesar Supplier No. 012609.A7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 584-08-7 | CK2O3 | 138.21 g/mol

It may be used in pharmaceutical laboratories as a drying agent or as a source of potassium. It is also used in fire extinguishers, to make soap, glass and to soften water. Aqueous potassium carbonate plays an important role in carbon dioxide absorption promoted by piperazine. It is also essential for the absorption of carbon dioxide promoted by boric acid. It is also useful for the synthesis of azoles and diazines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color Yellow
Physical Form Granules
Quantity 5 kg
Assay Percent Range 99.0% min.
Beilstein 4267587
Solubility Information Soluble in water. Insoluble in ethanol.
IUPAC Name dipotassium carbonate
Formula Weight 138.21
Grade ACS
Sensitivity Hygroscopic
Density 2.29 g/cm3 at 20°C
Chemical Name or Material Potassium carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Carbonic acid, dipotassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.