missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals

Product Code. 15454857
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454857 5 g 5g
15444857 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454857 Supplier Thermo Scientific Alfa Aesar Supplier No. H32168.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 888711-44-2 | Molecular Formula: CH2BBrF3K | Molecular Weight: 200.835 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 888711-44-2
Molecular Formula CH2BBrF3K
Molecular Weight (g/mol) 200.835
MDL Number MFCD09265154
InChI Key AZDFPIRYUOCVCJ-UHFFFAOYSA-N
Synonym potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v
PubChem CID 23690312
IUPAC Name potassium;bromomethyl(trifluoro)boranuide
SMILES [B-](CBr)(F)(F)F.[K+]

Specifications

Linear Formula BrCH2BF3K
Quantity 5 g
Formula Weight 200.84
Percent Purity 95%
Chemical Name or Material Potassium bromomethyltrifluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.