missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, ultra dry, 99.98% (metals basis)

Product Code. 11375757
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375757 500 g 500g
11365757 25 g 25g
11385757 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375757 Supplier Thermo Scientific Alfa Aesar Supplier No. 043448.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-02-3 | BrK | 119.00 g/mol

Potassium bromide is used in manufacturing of photographic papers, engraving and lithography, in spectroscopy and as an analytical lab reagent. Potassium bromide is employed in antiepileptic medication for dogs. It is utilized in the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Color White
Physical Form Beads
Quantity 500 g
Merck Index 14,7618
Solubility Information Freely soluble in water. Soluble in alcohol and glycerol. Slightly soluble in diethyl ether.
IUPAC Name potassium bromide
Formula Weight 119.01
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Refractive Index 1.559
Sensitivity Hygroscopic
Density 2.75 g/mL
Assay (metals basis)
Chemical Name or Material Potassium bromide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.