missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, TraceSELECT™, ≥99.99% (Metals Basis), Honeywell™ Fluka™

Product Code. 15627440 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15627440 25 g Poly Bottle 25g
15617440 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15627440 Supplier Honeywell-Fluka Supplier No. 9073725G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1383°C
Melting Point 734°C
Color Colorless
Quantity 25 g
Linear Formula KBr
UN Number NONH for all modes of transport
Solubility Information 656g/L at 20°C
Formula Weight 119g/mol
Percent Purity ≥99.9995% (metals basis)
Grade TraceSELECT
Odor Odorless
Packaging Poly Bottle
Appearance Solid
Vapor Pressure 0.01mmHg (20°C), 1mmHg (795°C)
Density In water 2.73g/cm3 at 20°C
Chemical Name or Material Potassium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.