missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, FTIR Grade

Product Code. 11359107
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359107 100 g 100g
11344757 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359107 Supplier Thermo Scientific Alfa Aesar Supplier No. 039794.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-02-3 | BrK | 119.00 g/mol

Potassium bromide is used as a veterinary drug, as an antiepileptic medication for dogs. It used for removal of peripheral membrane proteins. Potassium bromide and sodium perborate play an important role for mild and regioselective oxidative bromination of anilines. Its Pellet is useful for fine structural characteristics by infared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Physical Form Powder
Quantity 100 g
Solubility Information Soluble in water and slightly soluble in diethyl ether. Insoluble in acetate.
IUPAC Name potassium;bromide
Formula Weight 119.01
Grade Premium FTIR
Refractive Index 1.559
Sensitivity Hygroscopic
Density 2.75 g/mL
Chemical Name or Material Potassium Bromide, FTIR Grade
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.