missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, Certified AR for Analysis, Fisher Chemical™

Product Code. 10385950 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10385950 500 g 500g
10705151 250 g 250g
10676072 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10385950 Supplier Fisher Chemical Supplier No. P/4000/53
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 734°C
Color White
pH 5 to 8.8
Physical Form Solid
Quantity 500 g
IUPAC Name potassium;bromide
Formula Weight 119g/mol
Packaging Plastic powder jar
Vapor Pressure <0.01mmHg at 20°C
compliance-icons
Product Identifier
  • Potassium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.