missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate, TRC

Product Code. 30419441
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419441 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419441 Supplier Toronto Research Chemicals Supplier No. TRCP695025100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Bromate
CAS 7758-01-2
Quantity 100 mg
Molecular Formula Br O3 . K
Synonym Bromic acid','potassium salt (1:1), Bromic acid','potassium salt (8CI,9CI), Potassium bromate
InChI Formula InChI=1S/BrHO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1
SMILES [K+].[O-]Br(=O)=O
IUPAC Name potassium;bromate
Molecular Weight (g/mol) 167
Formula Weight 165.8668
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.