missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate solution, 0.0167M EU Pharmacopoeia, Fisher Chemical™

Product Code. 12930874 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12930874 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12930874 Supplier Fisher Chemical Supplier No. VL 035 1000
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Especificaciones

Packaging Bottle
CAS 1-2-7758
Molecular Formula BrKO3
Concentration 0.0167 M
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES [O-]Br(=O)=O.[K+]
IUPAC Name potassium;bromate
Molecular Weight (g/mol) 167.00
PubChem CID 23673461
Quantity 1000 mL
ChEBI CHEBI:38211
Grade Eu. Ph. Reference
Chemical Name or Material Potassium bromate solution
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Potassium bromate 0.0167M. Volumetric Solution European Pharmacopoeia Ref: 3004400
Palabra de advertencia
  • Danger
Clasificación
  • Carcinogenicity Category 1B
Indicaciones de peligro
  • H350-May cause cancer.
Consejos de prudencia
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.