missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromate/Potassium bromide, 0.1N Standardized Solution

Product Code. 11363697
Change view
Click to view available options
Quantity:
1 L
4 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363697 4 L 4 L
11353697 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363697 Supplier Thermo Scientific Alfa Aesar Supplier No. 035593.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KBrO3/KBr

Potassium bromate/Potassium bromide is used as an analytical reagent for volumetric analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula KBrO3/KBr
MDL Number MFCD00011359

Specifications

Odor Odorless
Physical Form Liquid
Color Colorless
Solubility Miscible with water
Quantity 4 L
Chemical Name or Material Potassium bromate/Potassium bromide
compliance-icons
Product Identifier
  • Potassium bromate/Potassium bromide, 0.1N Standardized Solution
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Potassium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.