missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromate concentrate, For 1L standard solution, 1/60 M KBrO3 (0.1N), Solstice

Product Code. 15605220
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15605220 6 x 1 Ea. Pack of 6
15695210 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15605220 Supplier Honeywell-Fluka Supplier No. 380806X1EA
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 1L standard solution, 1/60 M KBrO3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

CAS 7758-01-2, 7732-18-5
CAS Min % ≥3.0000%, ≥95.0000%
CAS Max % <5.0000%, ≤100.0000%
Physical Form Liquid
Quantity 6 x 1 Ea.
Packaging Ampoule
Density 1.024 g/cm3
Linear Formula KBrO3
UN Number NONH for all modes of transport
Formula Weight 167
Grade Not Available
Chemical Name or Material Potassium bromate concentrate, For 1L standard solution, 1/60 M KBrO3 (0.1N)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.