missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Borohydride, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Potassium Borohydride |
| CAS | 13762-51-1 |
| Molecular Formula | KBH4 |
| Quantity | 1 g |
| Synonym | Potassium Tetrahydroborate |
| InChI Formula | InChI=1S/BH4.K/h1H4;/q-1;+1 |
| SMILES | [BH4-].[K+] |
| IUPAC Name | potassium;boranuide |
| Molecular Weight (g/mol) | 53.94 |
| Formula Weight | 54.0043 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction