missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium borohydride, 98%, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
1 kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics
Specifications
Melting Point | >400°C (decomposition) |
Physical Form | Granules |
Quantity | 500 g |
Linear Formula | KBH4 |
UN Number | UN1870 |
Merck Index | 14,7616 |
Solubility Information | Soluble in water (19g/100g ); methanol (3;9g/100g ); 4:1 water:methanol mixture (13g/100g ). |
IUPAC Name | potassium boranuide |
Formula Weight | 53.94 |
Percent Purity | 98% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'