missing translation for 'onlineSavingsMsg'
Learn More

Potassium borohydride, 98%, Thermo Scientific™

Product Code. 15472777
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13762-51-1
Molecular Formula BH4K
Molecular Weight (g/mol) 53.94
MDL Number MFCD00011396
InChI Key ICRGAIPBTSPUEX-UHFFFAOYSA-N
Synonym potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
PubChem CID 22892188
SMILES [BH4-].[K+]

Specifications

Melting Point >400°C (decomposition)
Physical Form Granules
Quantity 100 g
Linear Formula KBH4
UN Number UN1870
Merck Index 14,7616
Solubility Information Soluble in water (19g/100g ); methanol (3;9g/100g ); 4:1 water:methanol mixture (13g/100g ).
IUPAC Name potassium boranuide
Formula Weight 53.94
Percent Purity 98%
Odor Odorless
Refractive Index 1.494
Sensitivity Moisture sensitive
Density 1.17 g/mL
Chemical Name or Material Potassium borohydride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'