missing translation for 'onlineSavingsMsg'
Learn More

Potassium bisulfate, Puriss., 98-102%, Honeywell

Product Code. 15671150 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Paper / Poly Bag
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15671150 25 kg Paper / Poly Bag 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15671150 Supplier Honeywell Research Chemicals Supplier No. 1260525KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7646-93-7
Quantity 25 kg
Assay Percent Range 98 to 102%
Molecular Formula HKO4S
MDL Number MFCD00011404
UN Number UN2509
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
SMILES [K+].OS([O-])(=O)=O
IUPAC Name potassium;hydrogen sulfate
Molecular Weight (g/mol) 136.16
PubChem CID 516920
Grade puriss.
Packaging Paper / Poly Bag
Chemical Name or Material Potassium bisulfate, Puriss., 98-102%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.