missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bis(trimethylsilyl)amide (95%), TRC

Product Code. 30390704
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390704 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390704 Supplier Toronto Research Chemicals Supplier No. TRCK999970250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Bis(trimethylsilyl)amide
CAS 40949-94-8
Molecular Formula C6 H18 K N Si2
Quantity 250 mg
Synonym Silanamine, 1,1,1-trimethyl-N-(trimethylsilyl)-, potassium salt (1:1) (ACI), Potassium, [bis(trimethylsilyl)amino]- (7CI), Silanamine, 1,1,1-trimethyl-N-(trimethylsilyl)-, potassium salt (9CI), Hexamethyldisilazane potassium salt, KHMDS, N,N-Bis(trimethylsilyl)amine potassium salt, Potassium bis(trimethylsilyl)amide, Potassium hexamethyldisilazamide, Potassium hexamethyldisilazane, Potassium hexamethyldisilazide, Potassium hexamethyldisilyl azide
InChI Formula InChI=1S/C6H18NSi2.K/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1
SMILES C[Si](C)(C)N([K])[Si](C)(C)C
IUPAC Name [bis(trimethylsilyl)amino]potassium
Molecular Weight (g/mol) 199.483
Formula Weight 199.061
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.