missing translation for 'onlineSavingsMsg'
Learn More

Potassium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Product Code. 15492797
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium bis(trifluoromethylsulfonyl)imide is used as Flavor & Fragrance intermediates. It is used as an intermediate in Industries such as agrochemical, dyestuff, pharmaceutical, syntheses material, chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90076-67-8
Molecular Formula C2F6KNO4S2
Molecular Weight (g/mol) 319.234
MDL Number MFCD06200829
InChI Key KVFIZLDWRFTUEM-UHFFFAOYSA-N
Synonym potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide
PubChem CID 11099217
IUPAC Name potassium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+]

Specifications

Melting Point 198°C to 203°C
Linear Formula (CF3SO2)2NK
Quantity 250 mg
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information It is soluble in water. 10mg/mL.
Formula Weight 319.24
Physical Form Solid
Chemical Name or Material Potassium bis (trifluoromethylsulfonyl)imide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'