missing translation for 'onlineSavingsMsg'
Learn More

Potassium Biphthalate, TRC

Product Code. 30425108
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425108 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425108 Supplier Toronto Research Chemicals Supplier No. TRCP69875825G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium hydrogen phthalate
CAS 877-24-7
Molecular Formula C8 H5 O4 . K
Quantity 25 g
Synonym Phthalic acid monopotassium, 1,2-Benzenedicarboxylic acid','potassium salt (1:1), 1,2-Benzenedicarboxylic acid','monopotassium salt (9CI), Phthalic acid','monopotassium salt (8CI), Hydrogen potassium phthalate, KAP, Monopotassium 1,2-benzenedicarboxylate, Monopotassium phthalate, Potassium acid phthalate, Potassium biphthalate, Potassium hydrogen phthalate
InChI Formula InChI=1S/C8H6O4.K/c9-7(10)5-3-1-2-4-6(5)8(11)12;/h1-4H,(H,9,10)(H,11,12);/q;+1/p-1
SMILES [K+].OC(=O)c1ccccc1C(=O)[O-]
IUPAC Name potassium;2-carboxybenzoate
Molecular Weight (g/mol) 204.22
Formula Weight 203.9825
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.