missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, ACS Reagent, Honeywell Fluka™

Product Code. p-7071921 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15622370 100 g 100g
15642370 500 g 500g
15632370 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15622370 Supplier Honeywell-Fluka Supplier No. 237205100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, powder, crystals or granules, 99.7%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Physical Form Powder, Crystals and/or Granules
Quantity 100 g
Linear Formula KHCO3
UN Number NONH for all modes of transport
Beilstein 4535309
Formula Weight 100.12g/mol
Percent Purity 99%
Grade ACS Reagent
Chemical Name or Material Potassium Bicarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.