missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, 99.7-100.5%, (ACS Reagent Grade), MP Biomedicals

Product Code. 11487832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Unit Size:
100g
1kg
5kg
Product Code. Quantity unitSize
11487832 5 kg 5kg
11467832 100 g 100g
11477832 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11487832

Brand: MP Biomedicals 0215255794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Quantity 5 kg
Assay Percent Range 99.7-100.5%
Formula Weight 100.12
Grade ACS Reagent
Chemical Name or Material Potassium Bicarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.