missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimonyl tartrate trihydrate, 99+%

Product Code. 10105363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10105363 2.5 kg Glass bottle 2.5kg
10174800 100 g Glass bottle 100g
10772841 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10105363 Supplier Thermo Scientific Acros Supplier No. 223800025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28300-74-5 | C8H10K2O15Sb2 | 667.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H10K2O15Sb2
Molecular Weight (g/mol) 667.87
MDL Number MFCD00167056
Synonym Antimonyl potassium tartrate sesquihydrate

Specifications

Melting Point 100.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99% min. (Iodometry)
Merck Index 15, 691
Solubility Information Solubility in water: 8.3%. Other solubilities: 1g/3mL boiling water, 1g/15mL glycerol, insoluble in alcohol
Formula Weight 667.87
Percent Purity ≥99%
Infrared Spectrum Authentic
Loss on Drying 2.7% max. (105°C)
Packaging Glass bottle
Chemical Name or Material Potassium antimonyl tartrate sesquihydrate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.