missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium antimonate trihydrate, 94+%, Thermo Scientific Chemicals
55.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Description
Potassium antimonyl trihydrate is used as a detecting agent for sodium. It is used as a mordant/ fixing agent in leather and textile dying industries. It is also used as a analytical reagent and flux additive for electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystals or Powder |
Quantity | 50 g |
Assay Percent Range | >94% |
UN Number | 1549 |
Synonym | Potassium Pyroantimonate, Potassium Hexahydroxyantimonate |
Solubility Information | Soluble in water and glycerine. Insoluble in ethanol. |
Formula Weight | 262.90 (208.85 Anhydrous) |
Appearance | White powder |
Chemical Name or Material | Potassium Antimonate Trihydrate |
Safety and Handling
Product Identifier
- Potassium antimonate trihydrate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P330-Rinse mouth.
- P391-Collect spillage.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'