missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimonate trihydrate, 94+%, Thermo Scientific Chemicals

Product Code. 11328328
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Potassium antimonyl trihydrate is used as a detecting agent for sodium. It is used as a mordant/ fixing agent in leather and textile dying industries. It is also used as a analytical reagent and flux additive for electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12208-13-8
Molecular Formula KSbO3·3H2O
MDL Number MFCD00150439

Specifications

Color White
Physical Form Crystals or Powder
Quantity 250 g
Assay Percent Range >94%
Synonym Potassium pyroantimonate, Potassium hexahydroxyantimonate
Solubility Information Soluble in water and glycerine. Insoluble in ethanol.
Formula Weight 262.90 (208.85 Anhydrous)
Appearance White powder
Chemical Name or Material Potassium antimonate trihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'