missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, TRC

Product Code. 30012725
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012725 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012725 Supplier Toronto Research Chemicals Supplier No. TRCP6987205G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 127-08-2
Molecular Formula C2 H3 O2 . K
Quantity 5 g
Synonym Potassium acetate, Acetic acid potassium salt (1:1), Acetic acid, potassium salt (8CI,9CI), Potassium acetate (6CI,7CI), Catacyst LB, Cryotech E 36, DPG 35, Dabco K 2008, Dabco K 2097, Dabco P 15, LCM 1, LK 25, P 15, PC 46, Polycat 46, Potassium ethanoate, Safeway KA-HOT
InChI Formula InChI=1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1
SMILES [K+].CC(=O)[O-]
IUPAC Name potassium;acetate
Molecular Weight (g/mol) 98.14
Formula Weight 97.977
Chemical Name or Material Potassium Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.