missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate hydrate, Puratronic™, 99.997% (metals basis)

Product Code. 11382068
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382068 5 g 5g
11372068 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382068 Supplier Thermo Scientific Alfa Aesar Supplier No. 012975.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 254966-93-3 | C2H3KO2 | 98.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 254966-93-3
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate hydrate, c2h3o2.k.h2o, potassium acetate hydrate, puratronic, potassium acetate hydrate, puratronic r metals basis
PubChem CID 23702041
SMILES [K+].CC([O-])=O

Specifications

Color White
Physical Form Crystalline
Quantity 5 g
IUPAC Name potassium acetate
Formula Weight 98.15 (Anhydrous)
Percent Purity 99.997%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Potassium acetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.