missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, Endotoxin Tested, Granular, Multi-Compendial, U.S.P., J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
Specifications
Specifications
| CAS | 127-08-2 |
| Physical Form | Granules |
| Quantity | 2.5 kg |
| Molecular Formula | C2H3KO2 |
| Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
| InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
| SMILES | CC(=O)[O-].[K+] |
| IUPAC Name | potassium;acetate |
| Molecular Weight (g/mol) | 98.142 |
| PubChem CID | 517044 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction