missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate, ACS, 99% min

Product Code. 11376798
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
5 x 2 kg
Unit Size:
100g
10kg
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376798 5 x 2 kg 10kg
11356798 100 g 100g
11346798 500 g 500g
11366798 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11376798 Supplier Thermo Scientific Alfa Aesar Supplier No. 013449.F7
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol

Potassium acetate is used as deicer to remove snow and ice from the surface and is considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive; as a buffer in biological research; as extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins thereby removal of proteins from DNA. It plays an important role for preserving tissues, fixation and mummification. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292°C
Physical Form Crystalline
Quantity 5 x 2 kg
Assay Percent Range 99% min
Beilstein 3595449
Solubility Information Soluble in water,alcohol and ammonia. Insoluble in acetone and ether.
Formula Weight 98.15
Grade ACS Reagent
Sensitivity Hygroscopic
Density 1.57 g/mL
Chemical Name or Material Potassium acetate, For ACS analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.