missing translation for 'onlineSavingsMsg'
Learn More

Melford Potassium Acetate

Product Code. 30018711
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018711 1 kg 1kg
30018712 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018711 Supplier Melford Supplier No. P420001000.0
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

In research laboratories, potassium acetate is utilized for various purposes.

In research laboratories, potassium acetate is utilized for various purposes. It is often employed in nucleic acid purification procedures, such as RNA extraction, where it helps precipitate nucleic acids from solution.

Potassium acetate is also used in protein purification techniques, such as ammonium sulfate precipitation, to selectively precipitate proteins from solution based on their solubility characteristics.

Furthermore, potassium acetate is utilized as a buffering agent in biochemical assays and cell culture media. Its buffering capacity helps maintain a stable pH environment, which is critical for many enzymatic reactions and cell culture processes

Specifications

Chemical Name or Material Potassium Acetate
CAS 127-08-2
Quantity 1 kg
Molecular Formula C2H3O2K
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.