missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-methyl-beta-styryltrifluoroborate, 95%

Product Code. 11436850
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436850 1 g 1g
11446850 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436850 Supplier Thermo Scientific Alfa Aesar Supplier No. L17973.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 219718-86-2 | C9H9BF3K | 224.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 219718-86-2
Molecular Formula C9H9BF3K
Molecular Weight (g/mol) 224.07
MDL Number MFCD02093982
InChI Key IHPTVBKDBKCIOO-UHFFFAOYSA-N
Synonym potassium 4-methyl-beta-styryltrifluoroborate, potassio 4-methylstyryl trifluoroboron v, potassium 4-methyl-?-styryltrifluoroborate, potassium 4-methyl-, a-styryltrifluoroborate, potassium 4-methyl-.beta.-styryltrifluoroborate, potassium trifluoro e-2-4-methylphenyl ethenyl boranuide
PubChem CID 23666371
IUPAC Name potassium;trifluoro-[(E)-2-(4-methylphenyl)ethenyl]boranuide
SMILES [K+].CC1=CC=C(C=C[B-](F)(F)F)C=C1

Specifications

Quantity 1 g
Beilstein 7783635
Formula Weight 224.08
Percent Purity 95%
Chemical Name or Material Potassium 4-methyl-beta-styryltrifluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.