missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-(ethylthio)phenyltrifluoroborate, 96%, Thermo Scientific™

Product Code. 15439427
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15439427

Brand: Thermo Scientific Alfa Aesar H52677.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850623-75-5
Molecular Formula C8H9BF3KS
Molecular Weight (g/mol) 244.124
MDL Number MFCD04115771
InChI Key BVLNLNRSQDZWIL-UHFFFAOYSA-N
Synonym potassium 4-ethylthiophenyl trifluoroborate, potassium 4-ethylthio phenyl trifluoroborate, potassium 4-ethylsulfanyl phenyl trifluoroboranuide, potassium 4-ethylthio phenyltrifluoroborate, potassium 4-ethylthio phenyl-trifluoroboranuide, potassium ion 4-ethylsulfanyl phenyl trifluoroboranuide, potassium 4-ethylsulfanylphenyl-tris fluoranyl boranuide, potassium 4-ethylsulfanyl phenyl trifluoro borate 1-
PubChem CID 44717235
IUPAC Name potassium;(4-ethylsulfanylphenyl)-trifluoroboranuide
SMILES [B-](C1=CC=C(C=C1)SCC)(F)(F)F.[K+]

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 244.12
Percent Purity 96%
Chemical Name or Material Potassium 4-(ethylthio)phenyltrifluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.