missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-acetylphenyltrifluoroborate, 97%, Thermo Scientific™

Catalog Number 15412218
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
252726-24-2
C8H7BF3KO
226.047
MFCD07784376
JRUXIRSHKHCTTK-UHFFFAOYSA-N
potassium 4-acetylphenyl trifluoroboranuide, potassium 4-acetylphenyltrifluoroborate, pubchem11433, trifluoro 4-acetylphenyl potassioboron v, potassium 4-acetylphenyltrifluoroborate, potassium 4-acetylphenyl trifluoro borate 1-, potassium 4-acetylphenyl trifluoroborate
23665673
potassium;(4-acetylphenyl)-trifluoroboranuide
[B-](C1=CC=C(C=C1)C(=O)C)(F)(F)F.[K+]
This item is not returnable. View return policy
252726-24-2
C8H7BF3KO
226.047
MFCD07784376
JRUXIRSHKHCTTK-UHFFFAOYSA-N
potassium 4-acetylphenyl trifluoroboranuide, potassium 4-acetylphenyltrifluoroborate, pubchem11433, trifluoro 4-acetylphenyl potassioboron v, potassium 4-acetylphenyltrifluoroborate, potassium 4-acetylphenyl trifluoro borate 1-, potassium 4-acetylphenyl trifluoroborate
23665673
potassium;(4-acetylphenyl)-trifluoroboranuide
[B-](C1=CC=C(C=C1)C(=O)C)(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 290°C (decomposition)
Color White
Physical Form Powder
Quantity 1 g
Formula Weight 226.05
Percent Purity 97%
Chemical Name or Material Potassium 4-acetylphenyltrifluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'