missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3-methoxyphenyltrifluoroborate, 98%, Thermo Scientific™

Product Code. 15469957
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469957 5 g 5g
15459957 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469957 Supplier Thermo Scientific Alfa Aesar Supplier No. H53200.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 438553-44-7
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.036
MDL Number MFCD04039954
InChI Key GKDCWEPJOSDGHY-UHFFFAOYSA-N
Synonym potassium 3-methoxyphenyl trifluoroborate, potassium trifluoro 3-methoxyphenyl borate, potassium 3-methoxyphenyltrifluoroborate, potassium 3-methoxypheny trifluoroborate, potassium trifluoro 3-methoxyphenyl boranuide, amtb102, trifluoro 3-methoxyphenyl potassioboron v, potassium trifluoro-3-methoxyphenyl boranuide, potassium trifluoro 3-methoxyphenyl borate 1-, potassium ion trifluoro 3-methoxyphenyl boranuide
PubChem CID 23697282
IUPAC Name potassium;trifluoro-(3-methoxyphenyl)boranuide
SMILES [B-](C1=CC(=CC=C1)OC)(F)(F)F.[K+]

Specifications

Melting Point 164°C to 168°C
Quantity 5 g
Formula Weight 214.04
Percent Purity 98%
Chemical Name or Material Potassium 3-methoxyphenyltrifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.