missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3,4-dichlorophenyltrifluoroborate, 96%, Thermo Scientific™

Product Code. 15449977
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449977 5 g 5g
15439977 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15449977 Supplier Thermo Scientific Alfa Aesar Supplier No. H53219.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850623-68-6
Molecular Formula C6H3BCl2F3K
Molecular Weight (g/mol) 252.894
MDL Number MFCD04115763
InChI Key RTZOKQYJVLRZFW-UHFFFAOYSA-N
Synonym potassium 3,4-dichlorophenyl trifluoroborate, potassium 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyltrifluoroborate, c6h3bcl2f3.k, potassium 3,4-dichlorophenyl-trifluoroboranuide, potassium ion 3,4-dichlorophenyl trifluoroboranuide, potassium 3,4-dichlorophenyl trifluoro borate 1-, potassium 3,4-dichlorophenyl-tris fluoranyl boranuide
PubChem CID 44717229
IUPAC Name potassium;(3,4-dichlorophenyl)-trifluoroboranuide
SMILES [B-](C1=CC(=C(C=C1)Cl)Cl)(F)(F)F.[K+]

Specifications

Melting Point 178°C to 180°C
Quantity 5 g
Formula Weight 252.9
Percent Purity 96%
Chemical Name or Material Potassium 3,4-dichlorophenyltrifluoroborate
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.