missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln., Thermo Scientific Chemicals

Catalog Number p-7050634
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
3164-85-0
C8H15KO2
182.30
MFCD00045896
ZUFQCVZBBNZMKD-UHFFFAOYNA-M
23669619
[K+].CCCCC(CC)C([O-])=O
This item is not returnable. View return policy
3164-85-0
C8H15KO2
182.30
MFCD00045896
ZUFQCVZBBNZMKD-UHFFFAOYNA-M
23669619
[K+].CCCCC(CC)C([O-])=O

Used to convert the tert-butylammmonium salt of clavulanic acid into potassium clavulanate or clavulanate potassium. Also used as a catalyst for polyurethane systems (foams) and for unsaturated polyester resin systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Characteristic
Linear Formula K[OOCCH(C2H5)C4H9]
Quantity 2 g
UN Number UN1993
Synonym potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p
Solubility Information Partly miscible with water.
IUPAC Name potassium;2-ethylhexanoate
Formula Weight 182.3
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 75% w/w soln.
Physical Form Viscous Liquid
Assay (metals basis)
Chemical Name or Material Potassium 2-ethylhexanoate
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'