missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln.

Product Code. 11333538
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333538 10 g 10g
11343538 2 g 2g
11323538 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11333538 Supplier Thermo Scientific Alfa Aesar Supplier No. 039163.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3164-85-0 | C8H15KO2 | 182.30 g/mol

Used to convert the tert-butylammmonium salt of clavulanic acid into potassium clavulanate or clavulanate potassium. Also used as a catalyst for polyurethane systems (foams) and for unsaturated polyester resin systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3164-85-0
Molecular Formula C8H15KO2
Molecular Weight (g/mol) 182.30
MDL Number MFCD00045896
InChI Key ZUFQCVZBBNZMKD-UHFFFAOYNA-M
PubChem CID 23669619
SMILES [K+].CCCCC(CC)C([O-])=O

Specifications

Odor Characteristic
Linear Formula K[OOCCH(C2H5)C4H9]
Quantity 10 g
UN Number UN1993
Synonym potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p
IUPAC Name potassium 2-ethylhexanoate
Formula Weight 182.3
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 75% w/w soln.
Physical Form Viscous Liquid
Assay (metals basis)
Chemical Name or Material Potassium 2-ethylhexanoate
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.