missing translation for 'onlineSavingsMsg'
Learn More

Potash, sulfurated, S 11.8% min as sulfide

Product Code. 11394308
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394308 100 g 100g
11384308 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394308 Supplier Thermo Scientific Alfa Aesar Supplier No. 041550.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mixture composed of potassium polysulfides and potassium thiosulfate | CAS: 39365-88-3 | K4O3S3 | 300.57 g/mol

Potash is used in black/brown patina (copper, brass, bronze). Its use include decorative color effects on copper, brass, bronze and nickel. Sulfurated potash has been used in the treatment of acne and other skin disorders usually in the form of a lotion with zinc sulfate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39365-88-3
Molecular Formula K4O3S3
Molecular Weight (g/mol) 300.57
MDL Number MFCD00064604
InChI Key YFKFGPGTVQHNEF-UHFFFAOYSA-L
Synonym potash, sulfurated, sulfurated potash, potash, sulfurated usp, liver of sulfur, 2k.k2s.so32, potash sulfurated liver of sulfur, dipotassium hypo potassium sulfide, potash, sulfurated, s as sulfide, dipotassium thiosulfate mixture with potassium sulfide, dipotassium thiosulfate mixture with potassium sulfide k2sx
PubChem CID 56842985
IUPAC Name tetrapotassium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;sulfanide
SMILES [S--].[K+].[K+].[K+].[K+].[O-]S([O-])(=O)=S

Specifications

Color Green
Physical Form Chunks
Quantity 100 g
Assay Percent Range S 11.8% min as sulfide
Solubility Information Soluble in water
Sensitivity Air sensitive; Hygroscopic
Appearance Green chunks
Chemical Name or Material Potash, Mixture composed of potassium polysulfides and potassium thiosulfate
compliance-icons compliance-icons
Product Identifier
  • Potash, sulfated
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H228-Flammable solid.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.