missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10581-1-AP-20UL
This item is not returnable.
View return policy
Description
POT1 is a component of the telomerase ribonucleoprotein (RNP) complex that is essential for the replication of chromosome termini. It is a component of the double-stranded telomeric DNA-binding TRF1 complex which is involved in the regulation of telomere length by cis-inhibition of telomerase. Pot1 also acts as a single-stranded telomeric DNA-binding protein and thus may act as a downstream effector of the TRF1 complex and may transduce information about telomere maintece and/or length to the telomere terminus. It binds to two or more telomeric single-stranded 5′-TTAGGG-3′ repeats (G-strand) and with high specificity to a minimal telomeric single-stranded 5′-TAGGGTTAG-3′ sequence. Its activity is TERT dependent but it does not increase TERT activity.Specifications
| POT1 | |
| Polyclonal | |
| Unconjugated | |
| POT1 | |
| hPot1, POT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 101185, 25913 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q91WC1, Q9NUX5 | |
| Pot1 | |
| POT1 Fusion Protein Ag0875 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction