missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POT1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-176V
This item is not returnable.
View return policy
Description
POT1 Polyclonal specifically detects POT1 in Human samples. It is validated for Western Blot.
Specifications
| POT1 | |
| Polyclonal | |
| Western Blot | |
| Q9NUX5 | |
| POT1 | |
| A synthetic peptide made to an internal portion of the human POT1 protein sequence (between residues 200-300). [UniProt# Q9NUX5] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 95% homology to monkey, 82% homology to rat, and 72% homology to mouse. | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DKFZp586D211, hPot1, POT1 protection of telomeres 1 homolog, POT1-like telomere end-binding protein, protection of telomeres 1 homolog (S. pombe), protection of telomeres protein 1 | |
| Rabbit | |
| 71.4 kDa | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, DNA replication Transcription Translation and Splicing | |
| 25913 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction